missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 Antibody (CL5634) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61420IR
This item is not returnable.
View return policy
Description
SOX4 Monoclonal antibody specifically detects SOX4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SOX4 | |
| Monoclonal | |
| DyLight 755 | |
| ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
| This antibody was developed using a synthetic peptide derived from Q06945, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Stem Cell Markers, Stem Cells | |
| 6659 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5634 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction