missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG4L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00 €
Specifications
| Antigen | SPAG4L |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SPAG4L Polyclonal specifically detects SPAG4L in Human samples. It is validated for Western Blot.Specifications
| SPAG4L | |
| Polyclonal | |
| Rabbit | |
| Human | |
| dJ726C3.1, MGC33594, Sad1 and UNC84 domain containing 5, SPAG4L, Sperm-associated antigen 4-like protein, SUN domain-containing protein 5, testis and spermatogenesis related gene 4, Testis and spermatogenesis-related gene 4 protein, TSARG4sperm associated antigen 4-like | |
| SUN5 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q8TC36 | |
| 140732 | |
| Synthetic peptides corresponding to SPAG4L(sperm associated antigen 4-like) The peptide sequence was selected from the N terminal of SPAG4L. Peptide sequence MPRSSRSPGDPGALLEDVAHNPRPRRIAQRGRNTSRMAEDTSPNMNDNIL. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title