missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPANX-D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68961
This item is not returnable.
View return policy
Description
SPANX-D Polyclonal antibody specifically detects SPANX-D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SPANX-D | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| CT11.3;CT11.4;dJ171K16.1;SPANXC;SPANX-C;SPANXD;SPANX-D;SPANXE;SPANX-E;Sperm protein associated with the nucleus on the X chromosome D | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TSPEELVNDHARKNRINPLQMEEEE | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 64648 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction