missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
368.00 € - 572.00 €
Specifications
| Antigen | SPARC |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407980
|
Novus Biologicals
NBP1-80972-25ul |
25ul |
368.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711394
|
Novus Biologicals
NBP1-80972 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPARC Polyclonal specifically detects SPARC in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SPARC | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| P09486 | |
| 6678 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VLVTLYERDEDNNLLTEKQKLRVKKIHENEKRLEAGDHPVELLARDFEKNYNMYIFPVHWQFGQLDQHPIDGYLSHTELAPLRAPLIPMEHCTTRFFETCDLDNDKYIALDEWAGCFGIKQKDIDKDL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 35 kDa |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| SPARC | |
| IgG | |
| Affinity Purified | |
| Specificity of human SPARC antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title