missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
292.00 € - 572.00 €
Specifications
| Antigen | SPATA18 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18441521
|
Novus Biologicals
NBP1-81984-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18299995
|
Novus Biologicals
NBP1-81984 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPATA18 Polyclonal specifically detects SPATA18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SPATA18 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8TC71 | |
| 132671 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AENLKRLVSNETLRTLQEKLDFWLKEYNTNTCDQNLNHCLELIEQVAKVQGQLFGILTAAAQEGGRNDGVETIKSR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 61 kDa |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ32906, Mieap, mitochondria-eating protein, spermatogenesis associated 18 homolog (rat), Spermatogenesis-associated protein 18, SPETEX1 | |
| SPATA18 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title