missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA2L Antibody (Z2P2D5*C8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00 € - 351.00 €
Specifications
| Antigen | SPATA2L |
|---|---|
| Clone | Z2P2D5*C8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626838
|
Novus Biologicals
NBP2-50306-0.025ml |
0.025 mL |
190.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676806
|
Novus Biologicals
NBP2-50306 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPATA2L Monoclonal antibody specifically detects SPATA2L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SPATA2L | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, Cytokines and Growth Factors | |
| PBS | |
| 124044 | |
| IgG1 κ | |
| Protein A purified |
| Z2P2D5*C8 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C16orf76, chromosome 16 open reading frame 76, MGC26885, SPATA2-like protein, spermatogenesis associated 2-like, spermatogenesis-associated protein 2-like protein, tamo | |
| Peptide Sequence - LLYNSPGARP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title