missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPC24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16578651
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16578651 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16578651 Supplier Invitrogen™ Supplier No. PA595812

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAAFRDIEEV SQGLLSLLGA NRAEAQQRRL LGRHEQVVER LLETQDGAEK QLREILTMEK EVAQSLLNAK EQVHQGGVEL QQLEAGLQEA GEEDTRLKAS LLQLTRELEE LKEIEADLER QEKEVDEDTT VTIPSAVYVA QLYHQVSKIE WDYECEPGMV KGIHHGPSVA QPIHLDSTQL SRKFISDYLW SLVDTEW; Positive Samples: Jurkat, HeLa, 293T; Cellular Location: Chromosome, Nucleus, centromere, kinetochore.

Acts as a component of the essential kinetochore-associated NDC80 complex, which is required for chromosome segregation and spindle checkpoint activity. Required for kinetochore integrity and the organization of stable microtubule binding sites in the outer plate of the kinetochore.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPC24
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SPC24
Gene Accession No. Q8NBT2, Q9D083
Gene Alias 2410030K01Rik; AV109292; hSpc24; kinetochore protein Spc24; SPBC24; SPC24; SPC24, NDC80 kinetochore complex component; SPC24, NDC80 kinetochore complex component, homolog; SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae); spindle pole body component 24 homolog
Gene Symbols SPC24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human SPC24 (NP_8723191).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147841, 67629
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.