missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Dihydrochloride, Fisher BioReagents™

Product Code. 10700350
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10700350 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10700350 Supplier Fisher BioReagents Supplier No. BP29571
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibits protein synthesis | CAS: 22189-32-8 | C14H36Cl2N2O12 | 495.34 g/mol

Inhibits protein synthesis

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spectinomycin Dihydrochloride
Type Spectinomycin Dihydrochloride
Physical Form Powder/Solid
CAS 22189-32-8
Melting Point 210°C
Source Streptomyces spectabilis
Ignition Residue < 1.0%
Packaging Amber Glass
Specific Rotation 15 to 21°
Water 16.0 to 20.0%
Concentration >603μg/mg
Quantity 1 g
Molecular Formula C14H36Cl2N2O12
MDL Number MFCD00150886
Merck Index 15, 8866
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
SMILES [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
IUPAC Name dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
Molecular Weight (g/mol) 495.34
PubChem CID 67108398
ChEBI CHEBI:9217
Formula Weight 495.3
Percent Purity >603 μg/mg
Show More Show Less
compliance-icons
Product Identifier
  • Spectinomycin pentahydrate dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.