missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin dihydrochloride, MedChemExpress

Product Code. 16470928
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470928 5 g 5g
16463786 25 g 25g
16402179 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16470928 Supplier Medchem Express Supplier No. HYB0438
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Spectinomycin dihydrochloride is a broad-spectrum antibiotic and inhibits the growth of a variety of gram-positive and gram-negative organisms. Spectinomycin dihydrochloride acts by selectively targeting to the bacterial ribosome and interrupting protein synthesis. Spectinomycin dihydrochloride is also a noncompetitive inhibitor of td intron RNA with an Ki value of 7.2 mM-.

Spectinomycin dihydrochloride is a broad-spectrum antibiotic and inhibits the growth of a variety of gram-positive and gram-negative organisms. Spectinomycin dihydrochloride acts by selectively targeting to the bacterial ribosome and interrupting protein synthesis. Spectinomycin dihydrochloride is also a noncompetitive inhibitor of td intron RNA with an Ki value of 7.2 mM-.

Specifications

Chemical Name or Material Spectinomycin dihydrochloride
Physical Form Solid
CAS 21736-83-4
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C14H26Cl2N2O7
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 100 mg/mL (246.75 mM)
SMILES CN[C@@H]([C@H]([C@@H](NC)[C@@H]1O)O)[C@]2([H])[C@]1([H])O[C@@](O[C@H](C)C3)([H])[C@](O)(C3=O)O2.Cl.Cl
Molecular Weight (g/mol) 405.27
Formula Weight 405.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.