missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin dihydrochloride pentahydrate, Cell Culture Grade

Product Code. 15480207
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15480207 Supplier Thermo Scientific Alfa Aesar Supplier No. J61820.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Spectinomycin dihydrochloride pentahydrate, CAS # 22189-32-8, is the hydrochloride salt form of spectinomycin, an aminocyclitol compound showing antibacterial and antimicrobial activities. | CAS: 22189-32-8 | C14H36Cl2N2O12 | 495.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

  • Spectinomycin dihydrochloride pentahydrate is derived from Streptomyces spectabilis and presents antibacterial and antimicrobial activities
  • Spectinomycin hydrochloride is a compound that inhibits the 30S ribosomal subunit of bacteria and interrupts protein synthesis and peptide elongation, leading to bacterial cell death

Application

  • Spectinomycin dihydrochloride pentahydrate is a compound that is active against gram-negative bacteria
  • In the laboratory, this compound is used in cell culture applications
  • It prevents contamination and can be used to select for resistant bacteria and plant cells containing the marker gene Spcr
  • It has been used to study respiratory tract infections caused by Pasteurella multocida and Mannheimia haemolytica
  • In genetic approaches, this compound can be used to amplify the plasmid copy number
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-32-8
Molecular Formula C14H36Cl2N2O12
Molecular Weight (g/mol) 495.34
MDL Number MFCD00150886
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
Synonym Spectinomycin dihydrochloride
PubChem CID 67108398
ChEBI CHEBI:9217
IUPAC Name dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
SMILES [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Type Spectinomycin Dihydrochloride Pentahydrate
Physical Form Powder
Color White
Quantity 25 g
Solubility Information Soluble in water;Soluble in water,methanol,dimethyl sulfoxide,ethanol and dimethyl formamide.
Formula Weight 495.35 (405.28 Anhydrous)
Grade Cell Culture
Chemical Name or Material Spectinomycin dihydrochloride pentahydrate
compliance-icons
Product Identifier
  • Spectinomycin dihydrochloride pentahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Spectinomycin hydrochloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.