missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta III Mouse, Clone: SPTBN2/1247, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6712-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
Spectrin beta III | |
Cell Surface | |
SPTBN2/1247 | |
Unconjugated | |
XP_016873664 | |
Mouse | |
246kDa | |
Protein A/G Purified | |
RUO | |
6712 | |
IgG3 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
SPTBN2 | |
Recombinant human SPTBN2 fragment (aa356-475) | |
HeLa cells. Pancreas or Liver. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |