missing translation for 'onlineSavingsMsg'
Learn More

Spectrometric Oil Standard D3-100: B, Mo, Zn @ 100 μg/g in Aviation Reference Oil, VHG™

Product Code. 30040542
Change view
Click to view available options
Quantity:
240 mL
Unit Size:
240mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040542 240 mL 240mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040542 Supplier VHG Labs Supplier No. VHG9150012830249
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Spectrometric Oil Standard D3-100: B, Mo, Zn @ 100 μg/g in Aviation Reference Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boron/Molybdenum/Zinc
CAS 7440-42-8, 7439-98-7, 7440-66-6
Methods RDE
Analyte or Component Names Boron/Molybdenum/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Aviation Oil
Quantity 240 mL
Concentration or Composition (by Analyte or Components) Boron 100 μg/g/Molybdenum 100 μg/g/Zinc 100 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.