missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrophotometry Didymium UV and Visible Wavelength Standard, 298 to 865 nm, Reagecon
Shop All Reagecon Products

Specifications
Specifications
| Packaging | Quartz glass cuvette, permanently sealed |
| Physical Form | Liquid |
| UN Number | 2920 |
| Type | Wavelength Standard |
| Quantity | 1 Cuvette |
Safety and Handling
Product Identifier
- didymium oxide solution UV & visible wavelength standard
- Danger
- Flammable liquid Category 3
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H226-Flammable liquid and vapour.
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : perchloric acid, praesodymium (III) oxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?