missing translation for 'onlineSavingsMsg'
Learn More

Spermine, MedChemExpress

Product Code. 16498806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498806 Supplier Medchem Express Supplier No. HYB1777
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Spermine (NSC 268508) functions directly as a free radical scabenger to protect DNA from free radical attack. Spermine has antiviral effects.

Spermine (NSC 268508) functions directly as a free radical scabenger to protect DNA from free radical attack. Spermine has antiviral effects.

Specifications

Chemical Name or Material Spermine
Physical Form Oil
CAS 71-44-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H26N4
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym NSC 268508 Neuridine
Solubility Information H2O : 125 mg/mL (617.77 mM; Need ultrasonic)
SMILES NCCCNCCCCNCCCN
Molecular Weight (g/mol) 202.34
Formula Weight 202.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.