missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPG3A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | SPG3A |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647701
|
Novus Biologicals
NBP2-93512-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648100
|
Novus Biologicals
NBP2-93512-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPG3A Polyclonal antibody specifically detects SPG3A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SPG3A | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| AD-FSP, atlastin GTPase 1, atlastin1, atlastin-1, Brain-specific GTP-binding protein, EC 3.6.5.-, FSP1, GBP3, GBP-3, GTP-binding protein 3, Guanine nucleotide-binding protein 3, guanylate-binding protein 3, Spastic paraplegia 3 protein A, spastic paraplegia 3A (autosomal dominant), SPG3, SPG3AhGBP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ATL1 (NP_056999.2). MAKNRRDRNSWGGFSEKTYEWSSEEEEPVKKAGPVQVLIVKDDHSFELDETALNRILLSEAVRDKEVVAVSVAGAFRKGKSFLMDFMLRYMYNQESVDWVGDYNEPLTGFSWRGGSERETTGIQIWSEIFLINKPDGKKVAVLLMDTQGTFDSQSTLRDSATVFALSTMISSIQVYNLSQNVQEDDLQHLQLFTEYGRLAMEETFLKPFQSLIFLVRDWSFPYEFSYGADGGAKFLEKRLKVSGNQHEELQNVRKHIHSCFTNISCFLLPHPGLKVATNP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 51062 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title