missing translation for 'onlineSavingsMsg'
Learn More

SPHK1 Rabbit Recombinant Antibody, Proteintech

Product Code. 30235486 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235486 20 μL 20µL
30234231 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235486 Supplier Proteintech Supplier No. 826091RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The SPHK1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SPHK1. This antibody recognizes human, mouse antigen. The SPHK1 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifications

Antigen SPHK1
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Recombinant Monoclonal
Clone 2K12
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols SPHK
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8877
Target Species Human, Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 42 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.