missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spi-B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98958-100ul
This item is not returnable.
View return policy
Description
Spi-B Polyclonal antibody specifically detects Spi-B in Human samples. It is validated for Immunoprecipitation
Specifications
| Spi-B | |
| Polyclonal | |
| Immunoprecipitation 4-8 uL/mg of lysate | |
| SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Spi-B. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 6689 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction