missing translation for 'onlineSavingsMsg'
Learn More

Spike Sample Analysis Standard I For CLP, SPEX CertiPrep™

Product Code. 11465968 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465968 125 mL 125mL
11475968 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465968 Supplier SPEX Certiprep Supplier No. SPIKE1

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Indeholder

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al200μg/mL
As200μg/mL
Ba200μg/mL
Se200μg/mL
Tl200μg/mL
Fe100μg/mL
Co50μg/mL
Mn50μg/mL
Ni50μg/mL
Pb50μg/mL
Sb50μg/mL
V50μg/mL
Zn50μg/mL
Cu25μg/mL
Cr20μg/mL
Ag5μg/mL
Be5μg/mL
Cd5μg/mL

Certificeringer

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Overholdelse

GHS/OSHA Complaint

Tekniske data

Chemical Name or Material ICP Spike Sample 1
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique GC-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Concentration or Composition (by Analyte or Components) Components in 5% HNO3/tr Tartaric Acid/tr HF:
Al    •200μg/mL
Sb    •50μg/mL
As    •200μg/mL
Ba    •200μg/mL
Be    •5μg/mL
Cd    •5μg/mL
Cr    •20μg/mL
Co    •50μg/mL
Cu    •25μg/mL
Fe    •100μg/mL
Pb    •50μg/mL
Mn    •50μg/mL
Ni    •50μg/mL
Se    •200μg/mL
Ag    •5μg/mL
Tl    •200μg/mL
V    •50μg/mL
Zn    •50μg/mL

Product intended for laboratory use only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.