missing translation for 'onlineSavingsMsg'
Learn More

Spike Sample Analysis Standard I For CLP, SPEX CertiPrep™

Product Code. 11475968 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475968 500 mL 500mL
11465968 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475968 Supplier SPEX Certiprep Supplier No. SPIKE1500
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al200μg/mL
As200μg/mL
Ba200μg/mL
Se200μg/mL
Tl200μg/mL
Fe100μg/mL
Co50μg/mL
Mn50μg/mL
Ni50μg/mL
Pb50μg/mL
Sb50μg/mL
V50μg/mL
Zn50μg/mL
Cu25μg/mL
Cr20μg/mL
Ag5μg/mL
Be5μg/mL
Cd5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material ICP Spike Sample 1
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique GC-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Components in 5% HNO3/tr Tartaric Acid/tr HF:
Al    •200μg/mL
Sb    •50μg/mL
As    •200μg/mL
Ba    •200μg/mL
Be    •5μg/mL
Cd    •5μg/mL
Cr    •20μg/mL
Co    •50μg/mL
Cu    •25μg/mL
Fe    •100μg/mL
Pb    •50μg/mL
Mn    •50μg/mL
Ni    •50μg/mL
Se    •200μg/mL
Ag    •5μg/mL
Tl    •200μg/mL
V    •50μg/mL
Zn    •50μg/mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.