missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINK5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94549-0.02ml
This item is not returnable.
View return policy
Description
SPINK5 Polyclonal antibody specifically detects SPINK5 in Human samples. It is validated for Western Blot
Specifications
| SPINK5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Kazal type 5, serine peptidase inhibitor, Kazal type 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 895-1094 of human SPINK5 (NP_001121170.1). ERKKKDEEKSSSKPSNNAKDQCRQVQNEAEDAKFRQPGRSLASVARMSTDECSEFRNYIRNNELICPRENDPVHGADGKFYTNKCYMCRAVFLTEALERAKLQEKPSHVRASQEEDSPDSFSSLDSEMCKDYRVLPRIGYLCPKDLKPVCGDDGQTYNNPCMLCHENLIRQTNTHIRSTGKCEESSTPGTTAASMPPSDE | |
| 0.02 mL | |
| Angiogenesis, Cardiovascular Biology, Cell Biology | |
| 11005 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction