missing translation for 'onlineSavingsMsg'
Learn More

Spinosyn D 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316604
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316604 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316604 Supplier Dr. Ehrenstorfer Supplier No. DREA16972840AL100
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spinosyn D
CAS 131929-63-0
Analyte or Component Names Spinosyn D
Shipping Condition +20°C
Molecular Formula C42 H67 N O10
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym A 83543D, Spinosad D, (2S,3aR,5aS,5bS,9S,13S,14R,16aS,16bS)-2-[(6-Deoxy-2,3,4-tri-O-methyl-α-L-mannopyranosyl)oxy]-13-[[(2R,5S,6R)-5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]oxy]-9-ethyl-2,3,3a,5a,5b,6,9,10,11,12,13,14,16a,16b-tetradecahydro-4,14-dimethyl-1H-as-indaceno[3,2-d]oxacyclododecin-7,15-dione
InChI Formula InChI=1S/C42H67NO10/c1-11-26-13-12-14-35(53-37-16-15-34(43(6)7)24(4)49-37)23(3)38(45)33-20-31-29(32(33)21-36(44)51-26)17-22(2)28-18-27(19-30(28)31)52-42-41(48-10)40(47-9)39(46-8)25(5)50-42/h17,20,23-32,34-35,37,39-42H,11-16,18-19,21H2,1-10H3/t23-,24-,25+,26+,27-,28+,29-,30-,31-,32+,34+,35+,37+,39+,40-,41-,42+/m1/s1
SMILES CC[C@H]1CCC[C@H](O[C@H]2CC[C@@H]([C@@H](C)O2)N(C)C)[C@@H](C)C(=O)C3=C[C@H]4[C@@H]5C[C@@H](C[C@H]5C(=C[C@H]4[C@@H]3CC(=O)O1)C)O[C@@H]6O[C@@H](C)[C@H](OC)[C@@H](OC)[C@H]6OC
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 745.98
Quantity 1 mL
Formula Weight 745.4765 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.