missing translation for 'onlineSavingsMsg'
Learn More

SPP-86, MedChemExpress

Product Code. 16497077
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16497077

Brand: Medchem Express HY110193

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SPP-86 is a potent and selective cell permeable inhibitor of RET tyrosine kinase, with an IC50 of 8 nM. SPP-86 inhibits RET-induced phosphatidylinositide 3-kinases (PI3K)/Akt and MAPK signaling, also inhibits RET-induced estrogen receptorα (ERα) phosphorylation in MCF7 cells.

SPP-86 is a potent and selective cell permeable inhibitor of RET tyrosine kinase, with an IC50 of 8 nM. SPP-86 inhibits RET-induced phosphatidylinositide 3-kinases (PI3K)/Akt and MAPK signaling, also inhibits RET-induced estrogen receptorα (ERα) phosphorylation in MCF7 cells.

Specifications

Chemical Name or Material SPP-86
Physical Form Solid
CAS 1357349-91-7
Color Light Nude
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H15N5
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 100 mg/mL (360.59 mM; Need ultrasonic)
SMILES CC(C)N1C2=NC=NC(N)=C2C(C#CC3=CC=CC=C3)=N1
Molecular Weight (g/mol) 277.32
Formula Weight 277.32
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.