Learn More
GVS SPR401 Elipse Integra Mask with A1P3 Filters for Organic Gases and Dust
GREENER_CHOICE
Product Code. 18489635
Change view
Click to view available options
Filter Type:
A1P3
Unit Size:
Each
Product Code. Filter Type unitSize
18489635 A1P3 Each
1 options
Product Code. 18489635 Supplier GVS Supplier No. SPR401
In Stock
Add to basket
Edge
Add to basket
This mask is recommended for organic gases and dust. Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
This mask is recommended for organic gases and dust, pesticides, painting, solvents into automotive, construction, agriculture and shipyard industry or repair.
  • Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
  • Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
Specifications
Color Blue
Disposable Reusable
Filter Type A1P3