Learn More
GVS SPR501 Elipse Mask with P3 Filters
Description
This mask is recommended for dust and fumes containing substances such as micro-organisms, marble, gypsum, titanium oxide, soapstone, rock wool, wood, detergents, textile fibers, spices, salt, animal feeds, etc..
- Protects against dust that can cause lung disease. In particular, protects against coal, silica, cotton, iron ore, graphite, kaolin, zinc, aluminium dusts.
- Protects against harmful dusts such as asbestos, bauxite, coal, silica, iron, and against toxic dusts such as manganese, lead and chromium.
- Extremely lightweight and compact to give the wearer as much comfort as possible.
- It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Filters last up to 5 years and can be easily replaced.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
- Certifications: Mask Body EN140: 1998.
- P3 RD Filters: EN143: 2000+A1: 2006.
- CE 2797
Specifications
Specifications
| Color | Blue |
| Disposable | Reusable |
| Filter Type | P3 |