Learn More
GVS SPR505 Elipse Mask with B1P3 Filters
GREENER_CHOICE
Product Code. 18499665
Change view
Click to view available options
Filter Type:
B1P3
Unit Size:
Each
Product Code. Filter Type unitSize
18499665 B1P3 Each
1 options
Product Code. 18499665 Supplier GVS Supplier No. SPR505
Only null left
Add to basket
Edge
Add to basket
This mask is recommended for inorganic gases and dust. Extremely lightweight and compact to give the wearer as much comfort as possible. It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
This mask is recommended for Inorganic gases and dust. manufacturing including iodine, chlorine or formaldehyde such as in insulation, industrial or consumer products, metal separation, microelectronics.
  • Extremely lightweight and compact to give the wearer as much comfort as possible.
  • It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
  • Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
  • Filters last up to 3 years and can be easily replaced.
  • Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
  • Certifications: Mask Body EN140: 1998. B1P3 R D Filters EN14387: 2004 + A1: 2008. CE 2797
Specifications
Color Blue
Disposable Reusable
Filter Type B1P3