missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRR1A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
225.00 € - 507.00 €
Specifications
| Antigen | SPRR1A |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665300
|
Novus Biologicals
NBP2-93464-0.02ml |
0.02 mL |
225.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648230
|
Novus Biologicals
NBP2-93464-0.1ml |
0.1 mL |
507.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPRR1A Polyclonal antibody specifically detects SPRR1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SPRR1A | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 6698 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 19 kDa pancornulin, cornifin A, cornifin-A, small proline-rich protein 1A, Small proline-rich protein IA, SPR-IA, SPRK | |
| A synthetic peptide corresponding to a sequence within amino acids 1-89 of human SPRR1A (NP_005978.2). CPSTVTPAPAQQKTKQK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title