missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY2 Recombinant Rabbit Monoclonal Antibody (24GB2525)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557263
This item is not returnable.
View return policy
Description
SPRY2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a protein belonging to the sprouty family. The encoded protein contains a carboxyl-terminal cysteine-rich domain essential for the inhibitory activity on receptor tyrosine kinase signaling proteins and is required for growth factor stimulated translocation of the protein to membrane ruffles. In primary dermal endothelial cells this gene is transiently upregulated in response to fibroblast growth factor two. This protein is indirectly involved in the non-cell autonomous inhibitory effect on fibroblast growth factor two signaling. The protein interacts with Cas-Br-M (murine) ectropic retroviral transforming sequence, and can function as a bimodal regulator of epidermal growth factor receptor/mitogen-activated protein kinase signaling. This protein may play a role in alveoli branching during lung development as shown by a similar mouse protein.
Specifications
| SPRY2 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43597 | |
| SPRY2 | |
| A synthesized peptide derived from human Sprouty 2 (aa 250-315). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2525 | |
| Unconjugated | |
| SPRY2 | |
| hSPRY2; IGAN3; MGC23039; protein sprouty homolog 2; sprouty 2; sprouty homolog 2; sprouty homolog 2 (Drosophila); sprouty RTK signaling antagonist 2; sprouty2; SPRY2; spry-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10253 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction