Learn More
Invitrogen™ SPRY2 Recombinant Rabbit Monoclonal Antibody (24GB2525)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557264
 View more versions of this product
Product Code. 30275665
  • 146.00 € / 20µL
Description
SPRY2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a protein belonging to the sprouty family. The encoded protein contains a carboxyl-terminal cysteine-rich domain essential for the inhibitory activity on receptor tyrosine kinase signaling proteins and is required for growth factor stimulated translocation of the protein to membrane ruffles. In primary dermal endothelial cells this gene is transiently upregulated in response to fibroblast growth factor two. This protein is indirectly involved in the non-cell autonomous inhibitory effect on fibroblast growth factor two signaling. The protein interacts with Cas-Br-M (murine) ectropic retroviral transforming sequence, and can function as a bimodal regulator of epidermal growth factor receptor/mitogen-activated protein kinase signaling. This protein may play a role in alveoli branching during lung development as shown by a similar mouse protein.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SPRY2
Recombinant Monoclonal
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43597
SPRY2
A synthesized peptide derived from human Sprouty 2 (aa 250-315).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2525
Unconjugated
SPRY2
hSPRY2; IGAN3; MGC23039; protein sprouty homolog 2; sprouty 2; sprouty homolog 2; sprouty homolog 2 (Drosophila); sprouty RTK signaling antagonist 2; sprouty2; SPRY2; spry-2
Rabbit
Affinity chromatography
RUO
10253
-20°C