missing translation for 'onlineSavingsMsg'
Learn More

SPT3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15577531
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15577531 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577531 Supplier Invitrogen Supplier No. PA536134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SPT3 Polyclonal Antibody

SPT3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SPT3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SUPT3H
Gene Accession No. O75486
Gene Alias Cdc5-like protein; D9509.12; Positive regulator of Ty transcription; Protein SPT3; SPT3; SPT3 homolog, SAGA and STAGA complex component; SPT3L; SPT3-like protein; suppressor of Ty 3 homolog; suppressor of Ty 3 homolog (S. cerevisiae); SUPT3H; Transcription initiation protein SPT3 homolog; transcriptional regulator SPT3; YDR392W
Gene Symbols SUPT3H
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 181-230 of human SPT3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 685697, 8464
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.