missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-104
This item is not returnable.
View return policy
Description
SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Knockout Validated, Knockdown Validated, Block/Neutralize.
Specifications
| SR-BI | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.1% Sodium Azide | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
| Rabbit | |
| 82 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
| 949 | |
| Human, Mouse, Rat, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Immunoassay, KnockDown, Neutralization | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Simple Western 1:100, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 2.5 - 5 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 2.5 - 5 ug/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 26865459), Proximity Ligation Assay reported in scientific literature (10.1016/j.jbc.2021.100828), Knockout Validated reported in scientific literature (PMID 31462534), KnockDown Validated, Block/Neutralize reported in scientific literature (PMID 24859737) | |
| Q61009 | |
| SCARB1 | |
| A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [UniProt# Q61009] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, rat, mink, hamster, primate, rabbit and the long form of human SR-BI. Reactivity with CHO cells reported in scientific literature (PMID: 12356718). | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction