missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SR-BI Monoclonal Antibody (OTI1F3)
GREENER_CHOICE

Product Code. 15975914
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15975914 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15975914 Supplier Invitrogen™ Supplier No. MA526320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SR-BI Monoclonal Antibody for ICC/IF, Western Blot

High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR-BI
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCARB1
Gene Accession No. Q8WTV0
Gene Alias AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII
Gene Symbols SCARB1
Host Species Mouse
Immunogen Full length human recombinant protein of SCARB1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 949
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.