missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRBD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | SRBD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631071
|
Novus Biologicals
NBP2-93887-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677031
|
Novus Biologicals
NBP2-93887-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SRBD1 Polyclonal antibody specifically detects SRBD1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SRBD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| FLJ10379, H_NH0576F01.1, S1 RNA binding domain 1, S1 RNA-binding domain-containing protein 1, WUGSC:H_NH0576F01.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 330-600 of human SRBD1 (NP_060549.4). LGLEGAARALLEKPGELSLLSYIRPDVKGLSTLQDIEIGVQHILADMIAKDKDTLDFIRNLCQKRHVCIQSSLAKVSSKKVNEKDVDKFLLYQHFSCNIRNIHHHQILAINRGENLKVLTVKVNISDGVKDEFCRWCIQNRWRPRSFARPELMKILYNSLNDSFKRLIYPLLCREFRAKLTSDAEKESVMMFGRNLRQLLLTSPVPGRTLMGVDPGYKHGCKLAIISPTSQILHTDVVYLHCGQGFREAEKIKTLLLNFNCSTVVIGNGTA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55133 | |
| IgG | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts