missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRM, Mouse anti-Human, Clone: 2C1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006723-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: LCCQGECQWLHLDLIKEMRQFCQSLFPVVAYAYCTIPTYPSGQIGFMLCSKNPSTNFQEPVQPLTQQQVAQMQLKYYNSDVHRAAFVLPEFARKALNDVSpecifications
| SRM | |
| Monoclonal | |
| Unconjugated | |
| spermidine synthase | |
| PAPT/SPDSY/SPS1/SRML1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2C1 | |
| In 1x PBS, pH 7.4 | |
| BC000309 | |
| SRM | |
| SRM (AAH00309, 203 a.a. ∼ 301 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6723 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction