missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRp55 Antibody (SR1629), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22494-100ul
This item is not returnable.
View return policy
Description
SRp55 Monoclonal antibody specifically detects SRp55 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| SRp55 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6431 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| SR1629 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| B52, FLJ08061, pre-mRNA splicing factor SRP55, Pre-mRNA-splicing factor SRP55, serine/arginine-rich splicing factor 6, SFRS6, Splicing factor, arginine/serine-rich 6MGC5045, splicing factor, arginine/serine-rich, 55 kDa, SR splicing factor 6, SRP55arginine/serine-rich splicing factor 6 | |
| A synthesized peptide derived from human SRp55 (Uniprot #: Q13247) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction