Learn More
Invitrogen™ SRSF6 Recombinant Rabbit Monoclonal Antibody (24GB1825)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557266
 View more versions of this product
Product Code. 30275390
  • 146.00 € / 20µL
Description
SRSF6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SRSF6
Recombinant Monoclonal
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
G3V6S8, Q13247, Q3TWW8
Srsf6
A synthesized peptide derived from human SRSF6 (aa 164-199).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1825
Unconjugated
Srsf6
1210001E11Rik; AI314910; AW146126; B52; epididymis secretory protein Li 91; HEL-S-91; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 6; MGC5045; pre-mRNA-splicing factor SRP55; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 6; serine/arginine-rich splicing factor 6-like protein; Sfrs6; splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; SRP55; SRP55-1; SRSF6
Rabbit
Affinity chromatography
RUO
362264, 6431, 67996
-20°C