missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (6B11B4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37339
This item is not returnable.
View return policy
Description
SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CD15/Lewis X | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| Mouse | |
| 59 kDa | |
| 0.1 mL | |
| Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers | |
| 2526 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 6B11B4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FUT4 | |
| Purified recombinant fragment of human FUT4 (AA: 199-302) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction