missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSNA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94036-0.02ml
This item is not returnable.
View return policy
Description
SSNA1 Polyclonal antibody specifically detects SSNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SSNA1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| NA-14, nuclear autoantigen of 14 kDa, Sjogren syndrome nuclear autoantigen 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-119 of human SSNA1 (NP_003722.2). MTQQGAALQNYNNELVKCIEELCQKREELCRQIQEEEDEKQRLQNEVRQLTEKLARVNENLARKIASRNEFDRTIAETEAAYLKILESSQTLLSVLKREAGNLTKATAPDQKSSGGRDS | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Immunology | |
| 8636 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction