missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSR240612, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
SSR240612 is a potent, and orally active specific non-peptide bradykinin B1 receptor antagonist, with Kis of 0.48 nM and 0.73 nM for B1 kinin receptors of human fibroblast MRC5 and HEK cells expressing human B1 receptors, 481 nM and 358 nM for B2 receptors of guinea pig ileum membranes and CHO cells expressing human B1 receptor, respectively.
Specifications
Specifications
| Chemical Name or Material | SSR240612 |
| CAS | 464930-42-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C42H53ClN4O7S |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (126.04 mM) |
| SMILES | O=S(N[C@@H](C1=CC=C2C(OCO2)=C1)CC(N[C@@H](C(N(C)C(C)C)=O)CC3=CC=C(CN([C@@H]4C)[C@H](CCC4)C)C=C3)=O)(C5=CC(C=CC(OC)=C6)=C6C=C5)=O.Cl[H] |
| Molecular Weight (g/mol) | 793.41 |
| Formula Weight | 793.41 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction