missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SSRP1 Antibody - N-terminal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30844-100ug
This item is not returnable.
View return policy
Description
SSRP1 Polyclonal antibody specifically detects SSRP1 in Human,Mouse samples. It is validated for Western Blot
Specifications
| SSRP1 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| Chromatin-specific transcription elongation factor 80 kDa subunit, cisplatin-DNA SSRP, facilitates chromatin remodeling 80 kDa subunit, Facilitates chromatin transcription complex 80 kDa subunit, Facilitates chromatin transcription complex subunit SSRP1, FACT, FACT complex subunit SSRP1, FACT80FACT 80 kDa subunit, FACTp80, high mobility group box, hSSRP1, Recombination signal sequence recognition protein 1, structure specific recognition protein 1, Structure-specific recognition protein 1, T160 | |
| Recombinant protein consisting of amino acids 1-329 of the human SSRP1 protein conjugated to KLH. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6749 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction