missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSSCA1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93170-0.02ml
This item is not returnable.
View return policy
Description
SSSCA1 Polyclonal antibody specifically detects SSSCA1 in Mouse samples. It is validated for Western Blot
Specifications
| SSSCA1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Autoantigen p27, centromeric autoantigen (27kD), p27, Sjoegren syndrome/scleroderma autoantigen 1, Sjogren syndrome/scleroderma autoantigen 1, Sjogren's syndrome/scleroderma autoantigen 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of human SSSCA1 (NP_006387.1). MALNGAEVDDFSWEPPTEAETKVLQARRERQDRISRLMGDYLLRGYRMLGETCADCGTILLQDKQRKIYCVACQELDSDVDKDNPALNAQAALSQAREHQLASASELPLGSRPAPQPPVPRPEHCEGAAAGLKAAQGPPAPAVPPNTDVMACTQTALLQKLTWASAELGSSTSLETSIQLCGLIRACAEALRSLQQLQH | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 10534 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction