Learn More
Invitrogen™ SSTR5 Recombinant Rabbit Monoclonal Antibody (HL2756)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556885
Product Code. 30275284
  • 500.00 € / 100µL
Description
SSTR5 Recombinant Monoclonal Antibody for Western Blot
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predominantly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSTR5
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P35346
Sstr5
Synthetic peptide encompassing a sequence within the Intracellular domain human of SSTR5. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL2756
Unconjugated
Sstr5
Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5
Rabbit
Protein A
RUO
6755
-20°C, Avoid Freeze/Thaw Cycles