Learn More
Invitrogen™ SSX2IP Recombinant Rabbit Monoclonal Antibody (24GB5255)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557273
 View more versions of this product
Product Code. 30275697
  • 500.00 € / 100µL
Description
SSX2IP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a protein that binds the cancer-testis antigen Synovial Sarcoma X breakpoint 2 protein. The encoded protein may regulate the activity of Synovial Sarcoma X breakpoint 2 protein in malignant cells. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SSX2IP
Recombinant Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8CGZ2, Q9Y2D8
SSX2IP
A synthesized peptide derived from human SSX2IP (aa 580-614).
100 μL
Primary
Human, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5255
Unconjugated
SSX2IP
ADIP; Afadin- and alpha-actinin-binding protein; afadin- and alpha-actinin-binding protein ADIP; Afadin- and alpha-actinin-binding protein-like protein; afadin DIL domain-interacting protein; AU014939; AU042321; FLJ10848; KIAA0923; LOW QUALITY PROTEIN: afadin- and alpha-actinin-binding protein; MGC75026; SSX family member 2 interacting protein; SSX2-interacting protein; Ssx2ip; synovial sarcoma, X breakpoint 2 interacting protein
Rabbit
Affinity chromatography
RUO
117178, 308023
-20°C