missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93153-0.1ml
This item is not returnable.
View return policy
Beskrivelse
SSX4 Polyclonal antibody specifically detects SSX4 in Human, Mouse samples. It is validated for Western Blot
Tekniske data
| SSX4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Cancer/testis antigen 5.4, CT5.4SSX4B, MGC119056, MGC12411, protein SSX4, SSX4A, synovial sarcoma, X breakpoint 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-188 of human SSX4 (NP_005627.1). FMRSKRAADFHGNDFGNDRNHRNQVERPQMTFGSLQRIFPKIMPKKPAEEENGLKEVPEASGPQNDGKQLCPPGNPSTLEKINKTSGPKRGKHAWTHRLRERKQLVVYEEISDPEEDDE | |
| 0.1 mL | |
| Cancer | |
| 6759 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion