missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST2/IL-33R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
369.00 € - 547.00 €
Specifications
| Antigen | ST2/IL-33R |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18410782
|
Novus Biologicals
NBP1-85251-25ul |
25ul |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18700754
|
Novus Biologicals
NBP1-85251 |
547.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ST2/IL-33R Polyclonal specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ST2/IL-33R | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed | |
| IL1RL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9173 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PDVLENKCGYTLCIYGRDMLPGEDVVTAVETNIRKSRRHIFILTPQITHNKEFAYEQEVALHCALIQNDAKVILIEMEALSELDMLQAEALQDSLQHLMKVQGTIKWREDHIANKRSLNSKFWKHVRYQMPVPSKIPRKASSLTPL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title