missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST3GAL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13546-1-AP-150UL
This item is not returnable.
View return policy
Description
Synthesis of alpha-2, 3-linked sialic acid to Gal(beta-1, 3)GalNAc is mediated by at least 3 distinct beta-galactoside alpha-2, 3-sialyltransferases (EC 2.4.99.4), including ST3GAL4. In contrast, only a single gene encodes the beta-galactoside alpha-2, 6-sialyltransferase, ST6GAL1.Specifications
| ST3GAL4 | |
| Polyclonal | |
| Unconjugated | |
| ST3GAL4 | |
| Alpha 2,3 sialyltransferase IV, Alpha 2,3 ST 4, CGS23, Gal NAc6S, NANTA3, SAT 3, SAT3, Sialyltransferase 4C, SIAT4, SIAT4 C, SIAT4C, ST 4, ST3Gal IV, ST3GAL4, ST3GalA.2, ST3GalIV, STZ | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20443, 363040, 6484 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61131, Q11206 | |
| St3gal4 | |
| ST3GAL4 Fusion Protein Ag3665 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction