missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST8SIA-VI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87042
This item is not returnable.
View return policy
Description
ST8SIA-VI Polyclonal specifically detects ST8SIA-VI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ST8SIA-VI | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| 8-sialyltransferase), alpha-2,8-sialyltransferase 8F, alpha-2,8-sialyltransferase 8F variant 2, SIA8F, sialytransferase St8Sia VI, SIAT8F, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6, ST8SiaVI, ST8SIA-VI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 338596 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ST8SIA6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RILVEESREATHGTPAALRTLRSPATAVPRATNSTYLNEKSLQLTEKCKNLQYGIESFSNKTKGYSENDYLQIITDIQSCPWKRQAEEYANFRAKLASCCDAVQNFVVSQNNTPVGTNMSYEVESKKEIPIKKNIFHMFPVSQP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction