missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Stackable Wire Rack for Enviro-Genie

Product Code. 10624653 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Enviro-Genie, Incubator-Genie
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10624653 Enviro-Genie, Incubator-Genie Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10624653 Supplier Scientific Industries Supplier No. 83041376
  • ONLINE EXCLUSIVE PRICE
    73.50 €
    62.34 € / Pack of 1
    Save 11.16 €
    15% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stackable Wire Rack is designed to increase shelf space in the temperature controlled chamber of the Enviro-Genie and the Incubator-Genie. Up to two racks can be added to the chamber.

  • Increases shelf space in the temperature controlled chamber of the Enviro-Genie or the Incubator-Genie
  • Two wire racks will fit inside the chamber to provide 3 total levels for containers
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stackable Rack
Material Metal
Weight (English) 2 lb.
Weight (Metric) 1 kg
Color Silver
For Use With (Equipment) Enviro-Genie, Incubator-Genie
No. of Shelves 1
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.