Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SF260plus, 15 mm
Product Code. 17598185 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF260plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17598185 SF260plus Sterilizer Each
1 options
Product Code. 17598185 Supplier Memmert™ Supplier No. SF260plusB29722
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SF260plus Sterilizer
Product Type Bottom Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel