missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless-Steel Seven Analysis Test Sieves, 200mm Dia.
Brand: RETSCH 60.322.000125
Additional Details : Weight : 0.00001kg
Specifications
Stainless Steel | |
200mm | |
1 Each | |
Test Sieve | |
25mm |
Stainless Steel | |
Separation, fractioning, particle size determination | |
125μm | |
ISO 3310 and ASTM E 11 |