Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN30plus, 15 mm
Product Code. 17548615 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN30plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17548615 SN30plus Sterilizer Each
1 options
Product Code. 17548615 Supplier Memmert™ Supplier No. SN30plusB02070
Only null left
Add to basket
Add to basket
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SN30plus Sterilizer
Product Type Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel